Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACCAGGATCTTCAGAACAAAGATG[G/T]TTTTTTGGGGGGCTTTGTCAGATGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
RIMKLA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RIMKLA - ribosomal modification protein rimK like family member A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZMYND12 - zinc finger MYND-type containing 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146192.1 | 1187 | Missense Mutation | AAC,ACC | N,T 196 | NP_001139664.1 | |
NM_032257.4 | 1187 | Missense Mutation | AAC,ACC | N,T 306 | NP_115633.3 |