Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATGGCTTTAAGGAATTCATAGCT[C/G]TCAAGATGACGTGATGCTATATGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604566 MIM: 605494 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALG6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ALG6 - ALG6, alpha-1,3-glucosyltransferase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ITGB3BP - integrin subunit beta 3 binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206739.1 | 654 | Silent Mutation | GAC,GAG | D,E 206 | NP_001193668.1 | |
NM_014288.4 | 654 | Silent Mutation | GAC,GAG | D,E 167 | NP_055103.3 | |
XM_006710514.3 | 654 | Intron | XP_006710577.3 | |||
XM_006710515.3 | 654 | Intron | XP_006710578.3 | |||
XM_017000853.1 | 654 | Intron | XP_016856342.1 | |||
XM_017000854.1 | 654 | Intron | XP_016856343.1 | |||
XM_017000855.1 | 654 | Silent Mutation | GAC,GAG | D,E 201 | XP_016856344.1 | |
XM_017000856.1 | 654 | Intron | XP_016856345.1 | |||
XM_017000857.1 | 654 | Intron | XP_016856346.1 |