Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCGGGAGCAGATCATGACCACCA[A/G]TGTCTGGCTGACCCAGGTAAGCGTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 118507 | ||||||||||||||||||||
Literature Links: |
CHRNB2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CHRNB2 - cholinergic receptor nicotinic beta 2 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000748.2 | 485 | Missense Mutation | AAT,AGT | N,S 80 | NP_000739.1 | |
XM_017000180.1 | 485 | UTR 5 | XP_016855669.1 |