Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAAGGTTCCATAGTCCTCAGCACC[C/T]CCCTGAGGAATTTGGCTTCGGCTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 605998 MIM: 616472 | ||||||||||||||||||||
Literature Links: |
HAX1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HAX1 - HCLS1 associated protein X-1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001018837.1 | Intron | NP_001018238.1 | ||||
NM_006118.3 | Intron | NP_006109.2 |
UBAP2L - ubiquitin associated protein 2 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |