Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCTGCCCTCTTTGACCACGCCCT[A/C]TTTGGGGAGGTGGCCTGCCGCCTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 139370 MIM: 606916 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GNAI3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GNAI3 - G protein subunit alpha i3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GPR61 - G protein-coupled receptor 61 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031936.4 | 3343 | Silent Mutation | CTA,CTC | L,L 109 | NP_114142.3 | |
XM_017002438.1 | 3343 | Silent Mutation | CTA,CTC | L,L 109 | XP_016857927.1 |