Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCGTGTAACGTGCGCTATGGAGC[C/G]GAAAGTCGCAGAGCTGAAGCAGAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
13 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 609831 MIM: 604746 | ||||||||||||||||||||
Literature Links: |
CCDC163P PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC163P - coiled-coil domain containing 163, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MMACHC - methylmalonic aciduria (cobalamin deficiency) cblC type, with homocystinuria | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015506.2 | 15 | Missense Mutation | CCG,CGG | P,R 3 | NP_056321.2 | |
XM_005270724.4 | 15 | Missense Mutation | CCG,CGG | P,R 3 | XP_005270781.1 | |
XM_011541204.2 | 15 | UTR 5 | XP_011539506.1 |
TESK2 - testis-specific kinase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |