Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGCAGAGGGAGGTCATGACTTTCA[C/G]TCTGCTGTTTAGCAAAATTTACAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 601691 | ||||||||||||||||||||
Literature Links: |
ABCA4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ABCA4 - ATP binding cassette subfamily A member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000350.2 | 6862 | Missense Mutation | ACT,AGT | T,S 2253 | NP_000341.2 |