Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGTGGGCTCTTCGGTAATACAGAG[C/G]GGGGAAGGTGTACTCAATTCCCAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
12 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138971 MIM: 611979 MIM: 608854 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CSF3R PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CSF3R - colony stimulating factor 3 receptor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MRPS15 - mitochondrial ribosomal protein S15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031280.3 | 741 | Missense Mutation | CCT,GCT | P,A 193 | NP_112570.2 |
OSCP1 - organic solute carrier partner 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |