Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCCTGGTTGTCATGTCCCATGAC[C/T]GCTATGTGGCCGTCTGCCGGTCCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
OR2W5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
OR2W5 - olfactory receptor family 2 subfamily W member 5 (gene/pseudogene) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004698.2 | 424 | Missense Mutation | CGC,TGC | R,C 122 | NP_001004698.1 |