Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCGCCTTCCTGCTCGACCTGGGCA[C/T]CGACCTGTGGGCCGCCGTCCAGTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SMPDL3B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SMPDL3B - sphingomyelin phosphodiesterase acid like 3B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009568.2 | 694 | Intron | NP_001009568.1 | |||
NM_001304579.1 | 694 | Intron | NP_001291508.1 | |||
NM_014474.3 | 694 | Intron | NP_055289.2 | |||
XM_011541259.2 | 694 | Intron | XP_011539561.1 | |||
XM_011541260.2 | 694 | Intron | XP_011539562.1 |
XKR8 - XK related 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018053.3 | 694 | Missense Mutation | ACC,ATC | T,I 29 | NP_060523.2 | |
XM_011541679.2 | 694 | UTR 5 | XP_011539981.1 | |||
XM_011541680.2 | 694 | UTR 5 | XP_011539982.1 |