Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGAGATGAAGATGATGATGAGGAA[C/G]AAGAAGAAGAAGGGGGCTCATGGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
9 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605998 MIM: 616472 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HAX1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HAX1 - HCLS1 associated protein X-1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001018837.1 | Intron | NP_001018238.1 | ||||
NM_006118.3 | Intron | NP_006109.2 |
UBAP2L - ubiquitin associated protein 2 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |