Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCAAACACTCACGTTGACCATAC[A/G]GTGAGTAATAAGATGCAGTCTTCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
12 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 609874 | ||||||||||||||||||||
Literature Links: |
ITLN2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ITLN2 - intelectin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080878.2 | 1242 | Missense Mutation | CCG,CTG | P,L 237 | NP_543154.1 | |
XM_011509210.1 | 1242 | Missense Mutation | CCG,CTG | P,L 237 | XP_011507512.1 | |
XM_011509211.1 | 1242 | Missense Mutation | CCG,CTG | P,L 236 | XP_011507513.1 |
LOC101928372 - uncharacterized LOC101928372 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |