Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGCGGCGAAAGCAGAGGGCAGCCC[G/T]CTTGTCACACTCGCAGGTCAGCCGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611758 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
OTUD3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OTUD3 - OTU deubiquitinase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PLA2G2E - phospholipase A2 group IIE | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014589.2 | 389 | Silent Mutation | AGG,CGG | R,R 111 | NP_055404.1 |