Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTATGAGCCAGAGAACATGGAAGC[C/T]GAGTGCTAGGAGCCCAGTGCCAAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610801 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC41A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC41A1 - solute carrier family 41 member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173854.5 | 2524 | Missense Mutation | AGC,GGC | S,G 497 | NP_776253.3 | |
XM_005245069.2 | 2524 | Missense Mutation | AGC,GGC | S,G 497 | XP_005245126.1 |