Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGAGCCCTGTGACATCATCGACAG[C/T]AGTGATGAGATGGATGCCCAGGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100506747 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC100506747 - alpha-1,3-mannosyl-glycoprotein 4-beta-N-acetylglucosaminyltransferase-like protein LOC641515 homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM183A - transmembrane protein 183A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322955.1 | 387 | Silent Mutation | AGC,AGT | S,S 96 | NP_001309884.1 | |
NM_001322956.1 | 387 | Silent Mutation | AGC,AGT | S,S 96 | NP_001309885.1 | |
NM_001322957.1 | 387 | Silent Mutation | AGC,AGT | S,S 96 | NP_001309886.1 | |
NM_001322958.1 | 387 | Silent Mutation | AGC,AGT | S,S 66 | NP_001309887.1 | |
NM_001322959.1 | 387 | Silent Mutation | AGC,AGT | S,S 66 | NP_001309888.1 | |
NM_138391.4 | 387 | Silent Mutation | AGC,AGT | S,S 96 | NP_612400.3 |