Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGTTCCATGCAGATGGCCCCTCC[A/G]CCCAGAACATACCTGCATGAGAAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TDRD10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TDRD10 - tudor domain containing 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UBE2Q1 - ubiquitin conjugating enzyme E2 Q1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017582.6 | 1129 | Silent Mutation | GGC,GGT | G,G 346 | NP_060052.3 |
UBE2Q1-AS1 - UBE2Q1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |