Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTCTGGTTACGGGGACACTGACGC[C/G]CGGGTCACCGCTGCCATAGCCAGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610389 MIM: 612942 MIM: 605440 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LAMTOR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LAMTOR2 - late endosomal/lysosomal adaptor, MAPK and MTOR activator 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145264.1 | 290 | Silent Mutation | GCC,GCG | A,A 42 | NP_001138736.1 | |
NM_014017.3 | 290 | Silent Mutation | GCC,GCG | A,A 42 | NP_054736.1 |
RAB25 - RAB25, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UBQLN4 - ubiquilin 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |