Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCGCCAGCTGCTGCTGAACTGCC[A/G]CCTGGTCTGCAGCCTCTGGCGGGAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 607112 MIM: 609111 MIM: 605647 | ||||||||||||||||||||
Literature Links: |
FBXO2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FBXO2 - F-box protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FBXO44 - F-box protein 44 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001014765.1 | 191 | Missense Mutation | CAC,CGC | H,R 31 | NP_001014765.1 | |
NM_001304790.1 | 191 | Missense Mutation | CAC,CGC | H,R 31 | NP_001291719.1 | |
NM_001304791.1 | 191 | Missense Mutation | CAC,CGC | H,R 31 | NP_001291720.1 | |
NM_033182.5 | 191 | Missense Mutation | CAC,CGC | H,R 31 | NP_149438.2 | |
NM_183412.2 | 191 | Missense Mutation | CAC,CGC | H,R 31 | NP_904319.1 | |
NM_183413.2 | 191 | Missense Mutation | CAC,CGC | H,R 31 | NP_904320.1 | |
XM_005263535.1 | 191 | Missense Mutation | CAC,CGC | H,R 31 | XP_005263592.1 | |
XM_005263536.4 | 191 | Missense Mutation | CAC,CGC | H,R 31 | XP_005263593.1 | |
XM_005263537.1 | 191 | Missense Mutation | CAC,CGC | H,R 31 | XP_005263594.1 | |
XM_006711043.2 | 191 | Missense Mutation | CAC,CGC | H,R 31 | XP_006711106.1 | |
XM_006711045.2 | 191 | Missense Mutation | CAC,CGC | H,R 31 | XP_006711108.1 | |
XM_011542435.1 | 191 | Missense Mutation | CAC,CGC | H,R 31 | XP_011540737.1 | |
XM_017002842.1 | 191 | Missense Mutation | CAC,CGC | H,R 31 | XP_016858331.1 | |
XM_017002843.1 | 191 | Missense Mutation | CAC,CGC | H,R 31 | XP_016858332.1 | |
XM_017002844.1 | 191 | Missense Mutation | CAC,CGC | H,R 31 | XP_016858333.1 |
FBXO6 - F-box protein 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |