Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTAGATCACTCCACTGGCCACTAC[A/G]AGTCCGGCACCTTCCCGGGCTGTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 614522 MIM: 603417 | ||||||||||||||||||||
Literature Links: |
KLHL12 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KLHL12 - kelch like family member 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303051.1 | Intron | NP_001289980.1 | ||||
NM_001303109.1 | Intron | NP_001290038.1 | ||||
NM_021633.3 | Intron | NP_067646.1 | ||||
XM_011509835.1 | Intron | XP_011508137.1 | ||||
XM_011509836.1 | Intron | XP_011508138.1 | ||||
XM_011509837.1 | Intron | XP_011508139.1 | ||||
XM_017001995.1 | Intron | XP_016857484.1 |
RABIF - RAB interacting factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |