Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTTGGCTTTGTGAACTCGCCTTC[A/G]AAGCTTCCTGCACATTCCATGTGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC190 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC190 - coiled-coil domain containing 190 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178550.4 | 3504 | Silent Mutation | NP_848645.3 | |||
XM_005245125.3 | 3504 | Silent Mutation | XP_005245182.1 | |||
XM_006711289.3 | 3504 | Silent Mutation | XP_006711352.1 | |||
XM_011509477.2 | 3504 | Silent Mutation | XP_011507779.1 |