Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAATGGGAAGGATACCTTTTGGGG[A/G]ATGATCCAGAGTGAACCAGAGCTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
12 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607945 MIM: 608341 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADIPOR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADIPOR1 - adiponectin receptor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CYB5R1 - cytochrome b5 reductase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016243.2 | 807 | Missense Mutation | NP_057327.2 |