Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCCTTCTTGCAGGTGCCGTGCTT[C/G]GCTCGGTGTCGTTGGAGGTGATTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF648 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF648 - zinc finger protein 648 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009992.1 | 5797 | Silent Mutation | GCC,GCG | A,A 552 | NP_001009992.1 | |
XM_017000301.1 | 5797 | Silent Mutation | GCC,GCG | A,A 552 | XP_016855790.1 | |
XM_017000302.1 | 5797 | Silent Mutation | GCC,GCG | A,A 552 | XP_016855791.1 |