Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCATGGCTGGAAGATCGACCTCCA[A/G]AAACATACAGAAGACCATTGACTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
8 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 147485 | ||||||||||||||||||||
Literature Links: |
IPP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IPP - intracisternal A particle-promoted polypeptide | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145349.1 | 1659 | Missense Mutation | TCT,TTT | S,F 538 | NP_001138821.1 | |
NM_005897.2 | 1659 | Missense Mutation | TCT,TTT | S,F 538 | NP_005888.1 | |
XM_005270839.4 | 1659 | UTR 3 | XP_005270896.1 | |||
XM_006710623.3 | 1659 | UTR 3 | XP_006710686.1 | |||
XM_011541394.2 | 1659 | Intron | XP_011539696.1 | |||
XM_017001210.1 | 1659 | Intron | XP_016856699.1 |
TMEM69 - transmembrane protein 69 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |