Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGCACAAGGATTAAAGGAAGCGT[C/T]AGTGGATACGCTCACCGGGATCCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611628 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IPO9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IPO9 - importin 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018085.4 | 124 | Missense Mutation | TCA,TTA | S,L 25 | NP_060555.2 |
IPO9-AS1 - IPO9 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NAV1 - neuron navigator 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |