Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGCCCACCGAAAGGCACCATCATT[C/T]CAGCCCAGCAGAAGTGTCCCTCAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 616363 | ||||||||||||||||||||
Literature Links: |
SPRR4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SPRR4 - small proline rich protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173080.2 | 942 | Missense Mutation | CCA,TCA | P,S 62 | NP_775103.1 | |
XM_017000482.1 | 942 | Missense Mutation | CCA,TCA | P,S 62 | XP_016855971.1 |