Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTTGTGCTGTTGCTGATGGATGGA[A/G]ATGTGGAAATGCTTCATGCAGGGAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 164850 | ||||||||||||||||||||
Literature Links: |
MYCL PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MYCL - v-myc avian myelocytomatosis viral oncogene lung carcinoma derived homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033081.2 | 1196 | Silent Mutation | NP_001028253.1 | |||
NM_001033082.2 | 1196 | Silent Mutation | NP_001028254.2 | |||
NM_005376.4 | 1196 | Intron | NP_005367.2 |