Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTCCCTTGTGCTTGTGTGGCAGG[G/T]TTGGTTTCAGAACAGAAGGGCCAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
9 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 616352 MIM: 602146 | ||||||||||||||||||||
Literature Links: |
ACBD6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACBD6 - acyl-CoA binding domain containing 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032360.3 | 1074 | Intron | NP_115736.1 |
LHX4 - LIM homeobox 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033343.3 | 1074 | Missense Mutation | GGT,GTT | G,V 203 | NP_203129.1 | |
XM_011510105.2 | 1074 | Missense Mutation | GGT,GTT | G,V 142 | XP_011508407.1 | |
XM_011510106.2 | 1074 | Missense Mutation | GGT,GTT | G,V 142 | XP_011508408.1 | |
XM_011510108.2 | 1074 | Missense Mutation | GGT,GTT | G,V 128 | XP_011508410.1 | |
XM_017002755.1 | 1074 | Missense Mutation | GGT,GTT | G,V 128 | XP_016858244.1 |
LHX4-AS1 - LHX4 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |