Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCATCAGCAATCAGAGAGGTCTA[A/G]ATCTCTGGAATCTGGCCACGGAGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 605051 MIM: 612280 | ||||||||||||||||||||
Literature Links: |
CNR2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CNR2 - cannabinoid receptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001841.2 | 1473 | Silent Mutation | CTA,TTA | L,L 355 | NP_001832.1 | |
XM_011540629.2 | 1473 | Silent Mutation | CTA,TTA | L,L 355 | XP_011538931.1 | |
XM_017000261.1 | 1473 | Silent Mutation | CTA,TTA | L,L 355 | XP_016855750.1 |
FUCA1 - fucosidase, alpha-L- 1, tissue | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |