Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCGGGGACGATGACATTGATGACG[C/T]TGTGGCCGCGCGCCAGTGTGGGGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601365 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DVL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DVL1 - dishevelled segment polarity protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MXRA8 - matrix remodeling associated 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282582.1 | 1264 | Missense Mutation | AAC,AGC | N,S 325 | NP_001269511.1 | |
NM_001282583.1 | 1264 | Missense Mutation | AAC,AGC | N,S 316 | NP_001269512.1 | |
NM_001282584.1 | 1264 | Missense Mutation | AAC,AGC | N,S 224 | NP_001269513.1 | |
NM_001282585.1 | 1264 | Missense Mutation | AAC,AGC | N,S 325 | NP_001269514.1 | |
NM_032348.3 | 1264 | Missense Mutation | AAC,AGC | N,S 325 | NP_115724.1 | |
XM_017001516.1 | 1264 | Missense Mutation | AAC,AGC | N,S 325 | XP_016857005.1 | |
XM_017001517.1 | 1264 | Missense Mutation | AAC,AGC | N,S 224 | XP_016857006.1 |