Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGATGGCCTGAAGTTTGGGCTGG[A/G]AGTGTAAGTAAAATATCGACATACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 613455 | ||||||||||||||||||||
Literature Links: |
MIA3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MIA3 - MIA family member 3, ER export factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300867.1 | 273 | Intron | NP_001287796.1 | |||
NM_001324062.1 | 273 | Silent Mutation | GGA,GGG | G,G 88 | NP_001310991.1 | |
NM_001324063.1 | 273 | Silent Mutation | GGA,GGG | G,G 88 | NP_001310992.1 | |
NM_001324064.1 | 273 | UTR 5 | NP_001310993.1 | |||
NM_001324065.1 | 273 | Intron | NP_001310994.1 | |||
NM_198551.3 | 273 | Silent Mutation | GGA,GGG | G,G 88 | NP_940953.2 | |
XM_006711304.3 | 273 | Silent Mutation | GGA,GGG | G,G 171 | XP_006711367.2 | |
XM_011509513.2 | 273 | Silent Mutation | GGA,GGG | G,G 171 | XP_011507815.2 | |
XM_017001243.1 | 273 | UTR 5 | XP_016856732.1 |