Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGAACAACCTGGGCAGGATCCCA[C/T]CTCAGACGACGTCATGGACTCGTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606753 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MROH7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MROH7 - maestro heat like repeat family member 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MROH7-TTC4 - MROH7-TTC4 readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TTC4 - tetratricopeptide repeat domain 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291333.1 | 113 | Missense Mutation | ACC,ATC | T,I 9 | NP_001278262.1 | |
NM_004623.4 | 113 | Missense Mutation | ACC,ATC | T,I 9 | NP_004614.3 |