Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGCACTCAGAACTGAGGCCGACA[C/T]CATCTCCGGGAGAGTGGATCGTCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608003 MIM: 610088 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HIPK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HIPK1 - homeodomain interacting protein kinase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OLFML3 - olfactomedin like 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286352.1 | 523 | Missense Mutation | ACC,ATC | T,I 62 | NP_001273281.1 | |
NM_001286353.1 | 523 | Missense Mutation | ACC,ATC | T,I 21 | NP_001273282.1 | |
NM_020190.3 | 523 | Missense Mutation | ACC,ATC | T,I 82 | NP_064575.1 | |
XM_017001848.1 | 523 | Missense Mutation | ACC,ATC | T,I 62 | XP_016857337.1 |