Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCACTGGGGCTTCGACTCGGCCA[A/G]CTGGCGGGCCTACATCCTGCTGAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 164780 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SKI PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SKI - SKI proto-oncogene | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003036.3 | 881 | Missense Mutation | AAC,AGC | N,S 280 | NP_003027.1 | |
XM_005244775.3 | 881 | Missense Mutation | AAC,AGC | N,S 280 | XP_005244832.1 | |
XM_005244776.4 | 881 | Intron | XP_005244833.1 | |||
XM_017002128.1 | 881 | Intron | XP_016857617.1 |