Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCTTGCTGCCCAGCTCCTCTTCA[A/G]TTCTTGGGAAGGAGAAAGGTAAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 172430 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ENO1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ENO1 - enolase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001201483.1 | 1760 | Missense Mutation | ACT,ATT | T,I 320 | NP_001188412.1 | |
NM_001428.3 | 1760 | Missense Mutation | ACT,ATT | T,I 413 | NP_001419.1 | |
XM_006710433.2 | 1760 | Missense Mutation | ACT,ATT | T,I 413 | XP_006710496.1 |
MIR6728 - microRNA 6728 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |