Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGGGCCCCCTGAGGGCCCCGAGA[C/T]GTTTGCTTTGTACATGGTGGACTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605548 MIM: 601380 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAM15 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADAM15 - ADAM metallopeptidase domain 15 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
EFNA4 - ephrin A4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005227.2 | 302 | Missense Mutation | ACG,ATG | T,M 72 | NP_005218.1 | |
NM_182689.1 | 302 | Missense Mutation | ACG,ATG | T,M 72 | NP_872631.1 | |
NM_182690.2 | 302 | Missense Mutation | ACG,ATG | T,M 72 | NP_872632.2 |
LOC100505666 - uncharacterized LOC100505666 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |