Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGTGTGGGGAGGTGCCCTGGGCCT[A/G]GGGCTGCTGCCTGTGCTGGCCTGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600241 MIM: 605875 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPR3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GPR3 - G protein-coupled receptor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005281.3 | 600 | Silent Mutation | CTA,CTG | L,L 167 | NP_005272.1 |
WASF2 - WAS protein family member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |