Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACATAAATCAATACTTACGTGCAA[C/G]CTTTGGATCACTGGGATTTTTCCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603341 MIM: 600024 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAH14 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNAH14 - dynein axonemal heavy chain 14 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LBR - lamin B receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002296.3 | 1562 | Missense Mutation | CTT,GTT | L,V 520 | NP_002287.2 | |
NM_194442.2 | 1562 | Missense Mutation | CTT,GTT | L,V 520 | NP_919424.1 | |
XM_005273125.2 | 1562 | Missense Mutation | CTT,GTT | L,V 478 | XP_005273182.1 | |
XM_011544185.2 | 1562 | Missense Mutation | CTT,GTT | L,V 520 | XP_011542487.1 |