Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTACTTGATTGAGGATACAGACA[C/T]GTGTTTCAGCTGGTTCTTGAGATCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 179836 | ||||||||||||||||||||
Literature Links: |
RPA2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RPA2 - replication protein A2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286076.1 | 1063 | Missense Mutation | ATG,GTG | M,V 141 | NP_001273005.1 | |
NM_001297558.1 | 1063 | Intron | NP_001284487.1 | |||
NM_002946.4 | 1063 | Missense Mutation | ATG,GTG | M,V 237 | NP_002937.1 |
THEMIS2 - thymocyte selection associated family member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |