Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGAGTCATCTCTGATGGATAACT[A/G]CTCAGAATTCTTCTCAATGCTTGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
14 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 600445 | ||||||||||||||||||||
Literature Links: |
ADORA3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADORA3 - adenosine A3 receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001302678.1 | 1786 | UTR 3 | NP_001289607.1 |
TMIGD3 - transmembrane and immunoglobulin domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001081976.2 | 1786 | Intron | NP_001075445.1 | |||
NM_001302680.1 | 1786 | Intron | NP_001289609.1 | |||
NM_020683.6 | 1786 | Intron | NP_065734.5 |