Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTACAGAGCCGTCTGGGTAGCGG[C/T]TCCTCCGCCTCGGTGTATCGGGTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608849 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
UHMK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
UHMK1 - U2AF homology motif (UHM) kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184763.1 | 292 | Intron | NP_001171692.1 | |||
NM_144624.2 | 292 | Silent Mutation | GGC,GGT | G,G 32 | NP_653225.2 | |
NM_175866.4 | 292 | Silent Mutation | GGC,GGT | G,G 32 | NP_787062.1 |