Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCAGGAGGAGGGAGTCTAGGGCT[G/T]AGGCTCCGCGTGGCCCTGTCGCTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602587 MIM: 614269 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACOT7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACOT7 - acyl-CoA thioesterase 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007274.3 | 1292 | Missense Mutation | AAG,CAG | K,Q 369 | NP_009205.3 | |
NM_181864.2 | 1292 | Missense Mutation | AAG,CAG | K,Q 379 | NP_863654.1 | |
NM_181865.2 | 1292 | Missense Mutation | AAG,CAG | K,Q 349 | NP_863655.1 | |
NM_181866.2 | 1292 | Missense Mutation | AAG,CAG | K,Q 328 | NP_863656.1 |
GPR153 - G protein-coupled receptor 153 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |