Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGATGAATTGCTAAGTGAAAAAC[C/T]AGAGGTCAAGCAGAAGTGGGCATCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
9 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 600727 | ||||||||||||||||||||
Literature Links: |
NFIA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NFIA - nuclear factor I A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134673.3 | 600 | Missense Mutation | CCA,CTA | P,L 65 | NP_001128145.1 | |
NM_001145511.1 | 600 | Missense Mutation | CCA,CTA | P,L 57 | NP_001138983.1 | |
NM_001145512.1 | 600 | Missense Mutation | CCA,CTA | P,L 110 | NP_001138984.1 | |
NM_005595.4 | 600 | Missense Mutation | CCA,CTA | P,L 65 | NP_005586.1 | |
XM_011541512.2 | 600 | Missense Mutation | CCA,CTA | P,L 65 | XP_011539814.1 | |
XM_011541514.2 | 600 | Missense Mutation | CCA,CTA | P,L 57 | XP_011539816.1 | |
XM_011541515.2 | 600 | Missense Mutation | CCA,CTA | P,L 65 | XP_011539817.1 | |
XM_017001362.1 | 600 | Missense Mutation | CCA,CTA | P,L 57 | XP_016856851.1 | |
XM_017001363.1 | 600 | Intron | XP_016856852.1 |