Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTACCTAAACCAGTCATATGGCATG[A/G]ACAATCACAGTGGTGGTGGTGGGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614601 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF326 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF326 - zinc finger protein 326 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320185.1 | 341 | Missense Mutation | AAC,GAC | N,D 60 | NP_001307114.1 | |
NM_181781.3 | 341 | Intron | NP_861446.2 | |||
NM_182975.3 | 341 | Missense Mutation | AAC,GAC | N,D 60 | NP_892020.1 | |
NM_182976.3 | 341 | Missense Mutation | AAC,GAC | N,D 60 | NP_892021.1 | |
XM_005270779.4 | 341 | UTR 5 | XP_005270836.1 | |||
XM_011541290.2 | 341 | UTR 5 | XP_011539592.1 |