Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGCCCAACAGGTAGAACTTTATGC[A/G]GAGCTCCAGGCCATGAACTTTGAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 602862 | ||||||||||||||||||||
Literature Links: |
UAP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
UAP1 - UDP-N-acetylglucosamine pyrophosphorylase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324113.1 | 385 | Silent Mutation | GCA,GCG | A,A 35 | NP_001311042.1 | |
NM_001324114.1 | 385 | Silent Mutation | GCA,GCG | A,A 35 | NP_001311043.1 | |
NM_001324115.1 | 385 | Silent Mutation | GCA,GCG | A,A 35 | NP_001311044.1 | |
NM_001324116.1 | 385 | Silent Mutation | GCA,GCG | A,A 35 | NP_001311045.1 | |
NM_001324117.1 | 385 | Silent Mutation | GCA,GCG | A,A 35 | NP_001311046.1 | |
NM_003115.4 | 385 | Silent Mutation | GCA,GCG | A,A 35 | NP_003106.3 | |
XM_011509913.2 | 385 | Silent Mutation | GCA,GCG | A,A 35 | XP_011508215.1 | |
XM_011509914.2 | 385 | Silent Mutation | GCA,GCG | A,A 35 | XP_011508216.1 |