Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATCTGGACCCAAAATGTTGGCCCC[C/T]GTTTGCCTGGTGGAAAATAACAATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612467 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GBP6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GBP6 - guanylate binding protein family member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320257.1 | 133 | Intron | NP_001307186.1 | |||
NM_198460.2 | 133 | Silent Mutation | CCC,CCT | P,P 10 | NP_940862.2 | |
XM_011540835.2 | 133 | Silent Mutation | CCC,CCT | P,P 10 | XP_011539137.1 |