Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGCATCCCCCTCCCGAGGGGTCA[A/G]ACAGAGAAGGCAGACCATGTGGATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606416 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NLRP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NLRP3 - NLR family pyrin domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079821.2 | 881 | Silent Mutation | CAA,CAG | Q,Q 45 | NP_001073289.1 | |
NM_001127461.2 | 881 | Silent Mutation | CAA,CAG | Q,Q 45 | NP_001120933.1 | |
NM_001127462.2 | 881 | Silent Mutation | CAA,CAG | Q,Q 45 | NP_001120934.1 | |
NM_001243133.1 | 881 | Silent Mutation | CAA,CAG | Q,Q 43 | NP_001230062.1 | |
NM_004895.4 | 881 | Silent Mutation | CAA,CAG | Q,Q 45 | NP_004886.3 | |
NM_183395.2 | 881 | Silent Mutation | CAA,CAG | Q,Q 45 | NP_899632.1 | |
XM_011544048.2 | 881 | Silent Mutation | CAA,CAG | Q,Q 45 | XP_011542350.1 | |
XM_011544053.2 | 881 | Silent Mutation | CAA,CAG | Q,Q 45 | XP_011542355.1 | |
XM_011544055.2 | 881 | Silent Mutation | CAA,CAG | Q,Q 45 | XP_011542357.1 | |
XM_017000181.1 | 881 | Silent Mutation | CAA,CAG | Q,Q 45 | XP_016855670.1 | |
XM_017000182.1 | 881 | Silent Mutation | CAA,CAG | Q,Q 45 | XP_016855671.1 | |
XM_017000183.1 | 881 | Silent Mutation | CAA,CAG | Q,Q 45 | XP_016855672.1 | |
XM_017000184.1 | 881 | Silent Mutation | CAA,CAG | Q,Q 45 | XP_016855673.1 |