Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTCACAGTCAAGAAAAATAAGAGA[C/T]ACATGGCTGAGGTGAATGCATCCCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 608507 | ||||||||||||||||||||
Literature Links: |
MFN2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MFN2 - mitofusin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127660.1 | 524 | Missense Mutation | CAC,TAC | H,Y 20 | NP_001121132.1 | |
NM_014874.3 | 524 | Missense Mutation | CAC,TAC | H,Y 20 | NP_055689.1 | |
XM_005263543.3 | 524 | Missense Mutation | CAC,TAC | H,Y 20 | XP_005263600.1 | |
XM_005263545.3 | 524 | Missense Mutation | CAC,TAC | H,Y 20 | XP_005263602.1 | |
XM_005263547.3 | 524 | Missense Mutation | CAC,TAC | H,Y 20 | XP_005263604.1 | |
XM_005263548.3 | 524 | Missense Mutation | CAC,TAC | H,Y 20 | XP_005263605.1 |