Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCATTCCCGGCGAGCTTTATCCT[A/G]TTCTTCTCGATGTCGTTTCCGACGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 116951 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDK11A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDK11A - cyclin-dependent kinase 11A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024011.2 | 493 | Nonsense Mutation | CAG,TAG | Q,* 139 | NP_076916.2 | |
NM_033529.2 | 493 | Nonsense Mutation | CAG,TAG | Q,* 129 | NP_277071.2 |
SLC35E2 - solute carrier family 35, member E2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |