Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCGGCGCTCTGCTTCGTCGTGGCC[A/G]TGGGCCTCGTGGCAGGCTGTGGCGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1orf210 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C1orf210 - chromosome 1 open reading frame 210 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM125 - transmembrane protein 125 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320244.1 | 856 | Missense Mutation | ATG,GTG | M,V 42 | NP_001307173.1 | |
NM_144626.2 | 856 | Missense Mutation | ATG,GTG | M,V 42 | NP_653227.1 | |
XM_005270467.3 | 856 | Missense Mutation | ATG,GTG | M,V 42 | XP_005270524.1 | |
XM_011540705.2 | 856 | Missense Mutation | ATG,GTG | M,V 42 | XP_011539007.1 |