Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGATCAGGAAGAGAGGGAGGTGAG[A/G]GAGGGACTGAGGGTTCTGAAGCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607646 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DCST2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DCST2 - DC-STAMP domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144622.2 | 1819 | Missense Mutation | CCC,CTC | P,L 759 | NP_653223.2 | |
XM_011509188.2 | 1819 | Missense Mutation | CCC,CTC | P,L 539 | XP_011507490.1 | |
XM_011509189.2 | 1819 | Missense Mutation | CCC,CTC | P,L 427 | XP_011507491.1 |
ZBTB7B - zinc finger and BTB domain containing 7B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252406.2 | 1819 | Intron | NP_001239335.1 | |||
NM_001256455.1 | 1819 | Intron | NP_001243384.1 | |||
XM_006711349.2 | 1819 | Intron | XP_006711412.2 | |||
XM_006711353.1 | 1819 | Intron | XP_006711416.1 | |||
XM_006711354.1 | 1819 | Intron | XP_006711417.1 | |||
XM_006711356.3 | 1819 | Intron | XP_006711419.1 | |||
XM_006711357.1 | 1819 | Intron | XP_006711420.1 | |||
XM_006711358.1 | 1819 | Intron | XP_006711421.1 | |||
XM_006711359.2 | 1819 | Intron | XP_006711422.1 | |||
XM_011509598.2 | 1819 | Intron | XP_011507900.1 | |||
XM_011509599.2 | 1819 | Intron | XP_011507901.1 | |||
XM_017001399.1 | 1819 | Intron | XP_016856888.1 |